missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534539
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is mouse brain tissue lysate. PA5-34539 can be used with blocking peptide PEP-1582.
KANK2 Polyclonal antibody specifically detects KANK2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
KANK2 | |
Polyclonal | |
Unconjugated | |
KANK2 | |
AI504612; ANKRD25; ankyrin repeat domain 25; ankyrin repeat domain-containing protein 25; BC010245; Kank2; KIAA1518; kidney ankyrin repeat-containing protein 2; KN motif and ankyrin repeat domain-containing protein 2; KN motif and ankyrin repeat domains 2; matrix-remodeling-associated protein 3; MXRA3; PPKWH; RGD1563532; SIP; SRC-1-interacting protein; SRC1-interacting protein; SRC-interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100361376, 235041, 25959 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q63ZY3, Q8BX02 | |
KANK2 | |
A 16 amino acid peptide near the carboxy terminus of human KANK2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |