missing translation for 'onlineSavingsMsg'
Learn More

JNK Mouse, PE, Clone: N9-66, BD

Product Code. 15861959
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15861959 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15861959 Supplier BD Biosciences Supplier No. 562480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The N9-66 monoclonal antibody specifically binds to JNK1 and JNK2 phosphorylated at the pT183/pY185 sites. c-Jun NH2-terminal Kinases (JNKs), also called Stress Activated Protein Kinases (SAPKs), are mitogen-activated protein kinases (MAPKs) with observed molecular weights of ∽46 kDa (JNK1) and ∽54 kDa (JNK2). Along with the p38 and ERK families, JNK represents one of three major classes of MAPKs. Complete activation of JNK requires the phosphorylation of both Thr183 and Tyr185 that are located in a Thr-X-Tyr motif. Phosphorylation of these residues is carried out by MKK4 and MKK7 that are phosphorylated and activated by MEKKs and MLKs in response to stress signals delivered through small GTPases of the Rho family. Once activated, JNK can translocate into the nucleus and regulate the expression of genes through phosphorylation of c-Jun, ATF-2, and other transcription factors. JNK plays a role in signal transduction in response to cytokines and various forms of environmental stress, such as endotoxins, UV irradiation, heat, and hyperosmolarity. JNK is critical to the regulation of cell growth, apoptosis, and the cellular response to stress, making it an important factor in tumorigenesis and adaptive immunity. During antibody development, the N9-66 monoclonal antibody was found to detect phosphorylated JNK1/2 by Western blot analysis of cellular lysates and by immunofluorescent staining and flow cytometric analysis of fixed and permeabilized cells. This antibody crossreacts with phosphorylated JNK1/2 expressed by mouse cells, as tested by Western blot analysis and flow cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK
Applications Flow Cytometry
Classification Monoclonal
Clone N9-66
Conjugate PE
Description MAPK8, MAPK9; SAPK1, SAPK; PRKM8, PRKM9
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated human JNK peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.