Learn More
IL-10 Rat anti-Human and Viral, Brilliant Violet 786, Clone: JES3-9D7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 564049
Description
The JES3-9D7 monoclonal antibody specifically reacts with human IL-10 (Interleukin-10) and viral IL-10. The immunogen used to generate the JES3-9D7 hybridoma was recombinant human IL-10 expressed by COS cells. IL-10 is also known as CSIF (Cytokine synthesis inhibitory factor) and (TGIF) T-cell growth inhibitory factor. IL-10 is expressed by various cell types including activated monocytes, macrophages, dendritic cells, mast cells, granulocytes, and lymphocytes. IL-10 is a pleiotropic cytokine that can downregulate proinflammatory immune responses, such as Th1-like responses, while promoting other responses including B cell proliferation and antibody production.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy7™-like dyes (eg, 780/60-nm filter).
Specifications
| IL-10 | |
| Monoclonal | |
| Brilliant Violet 786 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Recombinant Human IL-10 | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| JES3-9D7 | |
| CSIF; Cytokine synthesis inhibitory factor; TGIF | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human, Virus | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.