missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Mouse anti-Human, PE, Clone: G20-127, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562030
This item is not returnable.
View return policy
Description
The G20-127 monoclonal antibody specifically binds to the heavy chain of human IgM. The majority of mature B lymphocytes express IgM. The G20-127 antibody does not react with other immunoglobulin heavy chain isotypes.
Specifications
| IgM | |
| Monoclonal | |
| PE | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| G20-127 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction