missing translation for 'onlineSavingsMsg'
Learn More

IgG2a+b Rat anti-Mouse, PerCP, Clone: X57, BD

Product Code. 15805858
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15805858 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15805858 Supplier BD Biosciences Supplier No. 340271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Rat Anti-Mouse IgG2a+b, clone X57, is derived from hybridization of rat YB2/0 myeloma cells with spleen cells from LOU/M rats immunized with MOPC 141 mouse myeloma protein. (MOPC 141 is composed of mouse IgG2b heavy chains and kappa light chains.)

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a+b
Applications Flow Cytometry
Classification Monoclonal
Clone X57
Conjugate PerCP
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Rat
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.