missing translation for 'onlineSavingsMsg'
Learn More

IgG1 Rat anti-Mouse, PerCP, Clone: X56, BD

Product Code. 15815858
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15815858 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815858 Supplier BD Biosciences Supplier No. 340272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Rat Anti-Mouse IgG1, clone X56, is derived from hybridization of rat YB2/0 myeloma cells with spleen cells from LOU/M rats immunized with CD4 antibody. (CD4 is composed of mouse IgG1 heavy chains and kappa light chains.)

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Applications Flow Cytometry
Classification Monoclonal
Clone X56
Conjugate PerCP
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Rat
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.