missing translation for 'onlineSavingsMsg'
Learn More

Ig κ Light Chain Mouse anti-Human, APC, Clone: G20-193, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561323

Product Code. 15830949

  • ONLINE EXCLUSIVE PRICE
    195.00 €
    189.39 € / Each
    Save 5.61 €
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The G20-193 monoclonal antibody specifically binds to human immunoglobulin light chain, kappa (κ). It does not react with human immunoglobulin μ light chains or human immunoglobulin heavy chains.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ig κ Light Chain
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
G20-193
Ig, kappa Light Chain; IGKC; HCAK1; Km; Ig kappa chain C region
Mouse
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.