missing translation for 'onlineSavingsMsg'
Learn More

Ig Goat anti-Mouse, FITC, Polyclonal, BD

Product Code. 15826148
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15826148 500 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15826148 Supplier BD Biosciences Supplier No. 349031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Polyclonal anti-mouse Ig antibodies are obtained from antisera pooled from hyperimmunized goats.

The polyclonal antibodies are adsorbed against pooled human serum to remove any cross-reactivity to human Ig.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig
Applications Flow Cytometry
Classification Polyclonal
Clone POLYCLONAL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Goat
Quantity 500 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype Ig
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.