missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-γ Mouse anti-Human, Unlabeled, Clone: NIB42, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551221
This item is not returnable.
View return policy
Description
The NIB42 antibody reacts with human interferon-gamma (IFN-γ). The immunogen used to generate the NIB42 hybridoma was recombinant human IFN-γ. This is a neutralizing antibody.
Specifications
| IFN-γ | |
| Monoclonal | |
| 1.0mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant human IFN-γ | |
| 1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| ELISA | |
| NIB42 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction