Learn More
inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, Mouse, Clone: 3C3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ID2.
Brand: Abnova H00003398-M01.100ug
Description
The protein encoded by this gene belongs to the inhibitor of DNA binding (ID) family, members of which are transcriptional regulators that contain a helix-loop-helix (HLH) domain but not a basic domain. Members of the ID family inhibit the functions of basic helix-loop-helix transcription factors in a dominant-negative manner by suppressing their heterodimerization partners through the HLH domains. This protein may play a role in negatively regulating cell differentiation. A pseudogene has been identified for this gene. [provided by RefSeq
Sequence: MKAFSPVRSVRKNSLSDHSLGISRSKTPVDDPMSLLYNMNDCYSKLKELVPSIPQNKKVSKMEILQHVIDYILDLQIALDSHPTIVSLHHQRPGQNQASRTPLTTLNTDISILSLQASEFPSELMSNDSKALCGSpecifications
| inhibitor of DNA binding 2, dominant negative helix-loop-helix protein | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC030639 | |
| ID2 | |
| ID2 (AAH30639, 1 a.a. ∼ 134 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, KnockDown, Western Blot | |
| 3C3 | |
| Mouse monoclonal antibody raised against a full length recombinant ID2. | |
| ID2 | |
| GIG8/ID2A/ID2H/MGC26389/bHLHb26 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 3398 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.