Learn More
Anti-Human DPP4/CD26 (BA5b), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65267-1-IG-500UG
This item is not returnable.
View return policy
Description
The DPP4/CD26 antibody from Proteintech is a mouse monoclonal antibody generated with a human t cell clone of human DPP4/CD26. This antibody recognizes human antigen. The DPP4/CD26 antibody has been validated for the following applications: FC analysis.Specifications
| DPP4/CD26 | |
| Monoclonal | |
| 0.5 mg/mL | |
| ADABP; ADCP2; CD26; DPPIV; TP103 | |
| A human T cell clone | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| BA5b | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 1803 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |