missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-2Kk Monoclonal Antibody (H100-5-28), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517995
Additional Details : Weight : 0.01000kg
Description
MA5-17995 targets H-2Kk in FACS applications and shows reactivity with Mouse samples. The MA5-17995 immunogen is h-2Kk.
H-2Kk Monoclonal antibody specifically detects H-2Kk in Mouse samples. It is validated for Flow Cytometry.Specifications
H-2Kk | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P01901, P03991, P04223, P14428 | |
H2-K1 | |
H-2Kk | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
H100-5-28 | |
Biotin | |
H2-K1 | |
class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H2DD; H2K; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2K1; H2-K1; H2KD; H2KK; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen | |
Mouse | |
Protein G | |
RUO | |
14972 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |