missing translation for 'onlineSavingsMsg'
Learn More

GATA zinc finger domain containing 2B (A01), Mouse anti-Human, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16169106
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16169106 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16169106 Supplier Abnova Supplier No. H00057459A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant GATAD2B.

Sequence: RMTEDALRLNLLKRSLDPADERDDVLAKRLKMEGHEAMERLKMLALLKRKDLANLEVPHELPTKQDGSGVKGYEEKLNGNLRPHGDNRTAGRPGKENINDEPVDMSAR

Specifications

Antigen GATA zinc finger domain containing 2B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GATAD2B.
Formulation 50% glycerol
Gene GATAD2B
Gene Accession No. NM_020699
Gene Alias FLJ37346/KIAA1150/MGC138257/MGC138285/P66beta/RP11-216N14.6
Gene Symbols GATAD2B
Host Species Mouse
Immunogen GATAD2B (NP_065750, 3 a.a. ∼ 110 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 57459
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.