Learn More
GAD65 Mouse, Unlabeled, Clone: GAD-6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559931
Description
γ-aminobutyric acid (GABA) is an amino acid neurotransmitter that is considered to be the major inhibitory neurotransmitter in the mammalian central nervous system. The highest concentrations of GABA are found in the brain, where it is synthesized from glutamic acid to GABA by an enzyme called glutamic acid decarboxylase (GAD). GAD is also expressed in the insulin-producing β cells of the islets of Langerhans. Two isoforms of GAD are present in rat brain, GAD65 and GAD67, based on their relative molecular weight in kDa. Both isoforms have significant levels of homology in the catalytic portion of the molecule, but differ greatly in the first 95 amino acids in the N-terminal region. GAD65 migrates at ∽65 kDa in SDS/PAGE. The antibody is reported to recognize rat, human, mouse, and pig GAD65. Purified GAD65 from rat brain was used as the immunogen. The specific epitope recognized by this clone is a linear epitope localized in the last 41 amino acids of GAD65.
Specifications
| GAD65 | |
| Monoclonal | |
| 0.5mg/mL | |
| Glutamic acid decarboxylase | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse, Pig, Rat | |
| IgG2a |
| Western Blot | |
| GAD-6 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Purified Rat GAD65 | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.