Learn More
F3 (M01), Mouse anti-Human, Clone: 4G4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant F3.
Brand: Abnova H00002152-M01.100ug
Description
This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described. [provided by RefSeq
Sequence: TWKSTNFKTILEWEPKPVNQVYTVQISTKSGDWKSKCFYTTDTECDLTDEIVKDVKQTYLARVFSYPAGNVESTGSAGEPLYENSPEFTPYLETNLGQPTIQSFEQVGTKSpecifications
| F3 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| BC011029 | |
| F3 | |
| F3 (AAH11029, 45 a.a. ∼ 154 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 2152 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunoprecipitation, In situ PLA, Western Blot | |
| 4G4 | |
| Mouse monoclonal antibody raised against a partial recombinant F3. | |
| F3 | |
| CD142/TF/TFA | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.