missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin Mouse anti-Human, PE-CF594, Clone: CAM 5.2, BD

Product Code. 15812939
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15812939 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15812939 Supplier BD Biosciences Supplier No. 563615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CAM5.2 monoclonal antibody specifically recognizes cytokeratin having a primary reactivity with human keratin proteins that correspond to Moll's peptides #7 (48 kDa) and #8 (52 kDa). Cytokeratin 7 and 8 are type II cytoskeletal keratins. These cytoskeletal proteins provide structural integrity for epithelial cells and may serve other functions as well. They are expressed in epithelia cells that comprise normal human tissues. Although these cytokeratins are not normally expressed in stratified squamous epithelium, they may be expressed in some squamous cell carcinomas. The CAM 5.2 antibody stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin
Applications Flow Cytometry
Classification Monoclonal
Clone CAM 5.2
Conjugate PE-CF594
Description Cytokeratin-7/-8; CK-7/CK-8;
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HT29 colorectal carcinoma cell line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.