missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CR1, Mouse, Clone: 10D5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CX3CR1.
Brand: Abnova H00001524-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: MDQFPESVTENFEYDDLAEACYIGDIVVFGTVFLSISpecifications
| CX3CR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC028078 | |
| CX3CR1 | |
| CX3CR1 (AAH28078, 1 a.a. ∼ 36 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 10D5 | |
| Mouse monoclonal antibody raised against a partial recombinant CX3CR1. | |
| CX3CR1 | |
| CCRL1/CMKBRL1/CMKDR1/GPR13/GPRV28/V28 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1524 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- CX3CR1 monoclonal antibody (M07), clone 10D5
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction