missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CTDSP2 protein.
Brand: Abnova H00010106-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MEHGSIITQARREDALVLTKQGLVSKSSPKKPRGRNIFKALFCCFRAQHVGQSSSSTELAAYKEEANTIAKSDLLQCLQYQFYQIPGTCLLPEVTEEDQGRICVVIDLDETLVHSSFKPINNADFIVPIEIEGTTHQVYVLKRPYVDEFLRRMGELFECVLFTASLAKYADPVTDLLDRCGVFRARLFRESCVFHQGCYVKDLSRLGRDLRKTLILDNSPASYIFHPENAVPVQSWFDDMADTELLNLIPIFEELSGAEDVYTSLGQLRAPSpecifications
| CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human CTDSP2 protein. | |
| CTDSP2 | |
| OS4/PSR2/SCP2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10106 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005730.3 | |
| CTDSP2 | |
| CTDSP2 (NP_005721.3, 1 a.a. ∼ 271 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction