Learn More
Cryptosporidium parvum Oocysts Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA173184
Additional Details : Weight : 0.01000kg
Description
The PA1-73184 antibody reacts with Cryptosporidium parvum oocysts. PA1-73184 has been successfully used in immunofluorescence applications. The PA1-73184 antibody was raised against oocysts purified from bovine feces.
Cryptosporidium parvum Oocysts Polyclonal antibody specifically detects Cryptosporidium parvum Oocysts in Protozoa samples. It is validated for Immunocytochemistry.Specifications
Cryptosporidium parvum Oocysts | |
Polyclonal | |
FITC | |
Cryptosporidiosis | |
Intact oocysts purified from bovine feces. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide | |
Goat | |
purified | |
RUO | |
Protozoa | |
Antibody | |
IgG |