missing translation for 'onlineSavingsMsg'
Learn More

CrkL (pY207) Mouse, PE, Clone: K30-391.50.80, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560788

Product Code. 15800499

  • ONLINE EXCLUSIVE PRICE
    333.00 €
    331.70 € / Each
    Save 1.30 €
    0.39% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Crk-Like (CrkL) is an adaptor protein that is preferentially expressed in hematopoietic cells and is encoded by a gene that is homologous to the viral oncogene v-crk (chicken tumor virus no. 10 regulator of kinase). Its SH2 and SH3 domains bind to a variety of effector proteins, such as paxillin, p130Cas, c-Cbl, c-Abl, and C3G. These interactions are involved in the regulation of cellular migration, adhesion, and transformation. Tyrosine 207 (Y207) of CrkL is phosphorylated in hematopoietic cells that express the BCR-ABL fusion protein. This site
may be a negative regulator of protein complex formation and biological activity.

Intracellular Staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CrkL (pY207)
Monoclonal
PE
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
K30-391.50.80
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Phosphorylated Human, Mouse, and Rat CrkL Peptide
50 Tests
Cell Biology
Human, Mouse, Rat
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.