missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COLEC12 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547369
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse CL-P1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
COLEC12 Polyclonal antibody specifically detects COLEC12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Neutralization, Western Blot.Specifications
COLEC12 | |
Polyclonal | |
Unconjugated | |
Colec12 | |
CLP1; CL-P1; COLEC12; Collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; Scara4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; Srcl | |
Goat | |
Antigen affinity chromatography | |
RUO | |
81035 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q5KU26 | |
Colec12 | |
Mouse myeloma cell line NS0-derived recombinant human CL-P1/COLEC12 isoform 1 Leu57-Leu742 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |