missing translation for 'onlineSavingsMsg'
Learn More

Clostridium tetani Tetanus Toxin Mouse anti-Bacteria, Clone: 6E7, Invitrogen™

Product Code. 11544392
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11544392 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11544392 Supplier Invitrogen Supplier No. HYB2781702

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HYB 278-17-02 detects tetanus toxin and tetanus toxoid. HYB 278-17-02 has been successfully used in ELISA procedures. C. tetani toxoid injected in mice is neutralized by administration of HYB 278-17-02. The HYB 278-17-02 immunogen is Tetanus toxoid, formaldehyde inactivated Tetanus toxin. Epitope specificity of HYB 278-17-02 differs from that of HYB 278-14-02 and HYB 278-15-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL

Tetanus toxin is a neurotoxin produced by the vegetative spore of Clostridium tetani. The toxin causes tetanus, a disease that is characterized by prolonged contraction of skeletal muscle. World-wide, approximately 1 million cases of tetanus occurs annually, resulting in approximately 300,000 to 500,000 deaths, mostly in developing countries. The tetanus toxin has a molecular weight of approximately 150kDa and consists of a heavy chain (B-chain, 100kDa) and a light chain (A-chain, 50kDa) that are connected by a disulfide bond. The toxin acts by inhibiting neurotransmitter release. It binds to peripheral neuronal synapses, is internalized and moves by retrograde transport up the axon into the spinal cord where it can move between postsynaptic and presynaptic neurons. It inhibits the release of the neurotransmitters GABA and glycine by synaptobrevin-2. This results in overactivity in the muscle. The binding of the toxin to neurons is irreversible.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium tetani Tetanus Toxin
Applications ELISA
Classification Monoclonal
Clone 6E7
Concentration 1.06 mg/mL
Conjugate Unconjugated
Gene Alias C. tetani; Tetanus Toxin
Host Species Mouse
Immunogen Tetanus toxoid, formaldehyde inactivated Tetanus toxin
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.