Learn More
CLK3 (M07A), Mouse anti-Human, Clone: 7D6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CLK3.
Brand: Abnova H00001198-M07A.200uL
Description
This gene encodes a protein belonging to the serine/threonine type protein kinase family. This protein is a nuclear dual-specificity kinase that regulates the intranuclear distribution of the serine/arginine-rich (SR) family of splicing factors. Two transcript variants encoding different isoforms have been found for this gene. Related pseudogenes are located on chromosomes 1 and 9. [provided by RefSeq
Sequence: YPSRREPPPRRSRSRSHDRLPYQRRYRERRDSDTYRCEERSPSFGEDYYGPSRSRHRRRSRERGPYRTRKHAHHCHKRRTRSCSSASSRSQQSSKRSSRSVSpecifications
| CLK3 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC002555 | |
| CLK3 | |
| CLK3 (AAH02555, 36 a.a. ∼ 136 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1198 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 7D6 | |
| Mouse monoclonal antibody raised against a partial recombinant CLK3. | |
| CLK3 | |
| FLJ22858/PHCLK3/PHCLK3/152 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.