missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Mouse anti-Human, Clone: 23A1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0140
This item is not returnable.
View return policy
Description
A suggested positive control for this product is Bosc23 cells transfected with myc/His-CGA.
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.
Specifications
| Chromogranin A | |
| Monoclonal | |
| Unconjugated | |
| P10645 | |
| CHGA | |
| Recombit human CGA protein purified from E.coli | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 23A1 | |
| CHGA | |
| CGA | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1113 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction