missing translation for 'onlineSavingsMsg'
Learn More

CD91 Mouse anti-Human, PE, Clone: A2MR-A2, BD

Product Code. 15866288
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15866288 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15866288 Supplier BD Biosciences Supplier No. 550497

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reacts with a 600kDa, type I membrane single protein, also known as the α2 Macroglobulin (α2M) receptor/low density lipoprotein (LDL) receptor. The single chain receptor undergoes cleavage, shortly after synthesis, into the 85kDa transmembrane β chain that non-covalently binds to the extracellular 500-515kDa a chain. It has a broad cellular distribution, but in the hematopoietic system it is expressed on monocyte lineage cells. The α2M/LDL receptor mediates endocytosis of a variety of ligands including α2M-proteinase complexes, plasminogen activators in complex with plasminogen activator inhibitor, Pseudomonas Exotoxin A. Ligand binding to α2M/LDL receptor is followed by rapid transport of the ligand to lysosomes for degradation.

Host Species: Mouse
Clone: A2MR-α2
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD91
Applications Flow Cytometry
Classification Monoclonal
Clone A2MR-A2
Conjugate PE
Description Alpha-2-macroglobulin receptor; α2MR; LRP-1; LDL Receptor-Related Protein-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.