missing translation for 'onlineSavingsMsg'
Learn More

anti-CD8; Clone: RPA-T8; BD

Brand:  BD Biosciences 563256

Product Code. 15802649

  • ONLINE EXCLUSIVE PRICE
    263.00 €
    262.15 € / Each
    Save 0.85 €
    0.32% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Species: Mouse
Clone RPA-T8
Isotype IgG1, κ
Species Reactivity Hu, Rh, Cyn, Bb

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD8
RPA-T8
Mouse
50 Tests
Primary
IgG1
Monoclonal
Brilliant Violet 510
Affinity Purified
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.