Learn More
anti-CD79B, Human, Matched Antibody Pair, Abnova™
Product Code. 16121874
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16121874 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16121874 Supplier Abnova Supplier No. H00000974AP52.1Set
Please to purchase this item. Need a web account? Register with us today!
Includes
Capture antibody: mouse monoclonal anti-CD79B, IgG1 Kappa (300μg) , Detection antibody: rabbit polyclonal anti-CD79B (50μL) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen CD79B
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD79B.
Gene CD79B
Gene Alias B29/IGB
Gene Symbols CD79B
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 974
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD79B, IgG1 Kappa (300 ug) 2. Detection antibody: rabbit polyclonal anti-CD79B (50 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid