missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD63, PE; Clone: NVG-2; BD
Brand: BD Biosciences 564222
This item is not returnable.
View return policy
Description
Applications:
Flow cytometry, Intracellular staining
Specifications
| CD63 | |
| Monoclonal | |
| 0.2mg/mL | |
| Rat | |
| Affinity Purified | |
| Primary | |
| Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze |
| Flow Cytometry, Western Blot | |
| NVG-2 | |
| PE | |
| Mouse Eosinophils | |
| RUO | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction