missing translation for 'onlineSavingsMsg'
Learn More

anti-CD279 (PD-1); Clone: EH12.1; BD

Product Code. 15813099
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15813099 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15813099 Supplier BD Biosciences Supplier No. 563789

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Host Species: Mouse
Clone: EH12.1
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human PD-1 Recombinant Protein

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD279 (PD-1)
Classification Monoclonal
Clone EH12.1
Host Species Mouse
Immunogen Human PD-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.