Learn More
CD26 Mouse anti-Human, PE, Clone: L272, BD
Product Code. 15835878
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15835878 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15835878 Supplier BD Biosciences Supplier No. 340423
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
CD26, clone L272, is derived from hybridization of mouse Sp2/0 cells with spleen cells from BALB/c mice immunized with E1αPGF/JY cells.
CD26 recognizes a 120-kdalton (kDa) antigen that is identical to the enzyme dipeptidyl peptidase (DPP), a serine protease. It is associated with the binding of the TAT transactivating protein of the human immunodeficiency virus (HIV). CD26 and CD45 acts in a co-stimulatory fashion on T lymphocytes.
TRUSTED_SUSTAINABILITY
Specifications
Antigen CD26
Applications Flow Cytometry
Classification Monoclonal
Clone L272
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen E1aPGF/JY cells
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2a κ