Learn More
CD222 Mouse anti-Human, Alexa Fluor 647, Clone: MEM-238, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565105
Description
The MEM-238 monoclonal antibody specifically binds to CD222, which is also known as the Insulin-like growth factor 2 Receptor (IGF2R, IGF-II Receptor), Cation-independent mannose 6-phosphate receptor (CIMPR), or Mannose-6 phosphate receptor (M6PR). CD222 is ubiquitously expressed by a variety of cell types as a cell surface type I transmembrane glycoprotein. However, in the course of receptor trafficking between the cell surface and intracellular compartments, the majority of CD222 is found within cells. Cell surface CD222 functions as a multifunctional receptor that binds to a large number of extracellular ligands including acid hydrolases, insulin-like growth factors, latent TGF- β, leukemia inhibitory factor (LIF), proliferin, prorenin, plasminogen, and Herpes simplex vir us. It regulates extracellular Insulin-like growth factor II (IGF-II/IGF-2) levels by binding and internalizing the growth factor for lysosomal degradation. This effectively removes IGF-II from the circulation and tissues and thus prevents it from signaling through the growth-stimulatory Insulin-like growth factor I Receptor (IGF-1R, CD221) pathway. However, studies have reported that IGF-II may activate some cellular functions through CD222 as well. A soluble form of the CD222 extracellular region can also be detected in human serum and may play a role in regulating IGF-II activity. CD222 serves as a surface receptor for latent TGFβ and can complex with plasminogen and CD87, a urokinase-type plasminogen activator receptor, to activate latent TGF- β. CD222 also binds to a variety of Mannose 6-phosphate (M6P)-containing proteins, including extracellular and newly-synthesized lysosomal enzymes, and transports them to lysosomes.
Specifications
| CD222 | |
| Monoclonal | |
| 0.2mg/mL | |
| IGF2R; IGF-IIR; IGF-II Receptor; CIMPR; M6PR; MPR1; MPRI; M6P-R | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| MEM-238 | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing ≤0.09% sodium azide | |
| Human Recombinant Vaccinia virus encoding CD222 | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.