Learn More
CD10 Mouse anti-Human, FITC, Clone: W8E7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 347503
Description
The CD10 antibody, clone W8E7, is derived from the hybridization of P3-X63-Ag8.653 mouse myeloma cells with spleen cells of BALB/c mice immunized with blasts from a patient with non-T ALL.
The CD10 antibody (Anti-CALLA) recognizes a human common acute lymphoblastic leukemia antigen (CALLA), with a molecular weight of 100 kilodaltons (kDa). The CD10 antigen is identical to human membrane–associated neutral endopeptidase (NEP; EC 3.3.24.11), also known as enkephalinase.
Specifications
| CD10 | |
| Monoclonal | |
| 6μg/mL | |
| CALLA | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| W8E7 | |
| FITC | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human T-acute lymphoblastic leukemia (ALL) cells | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.