missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5R1, Mouse, Clone: 4E2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant C5R1.
Brand: Abnova H00000728-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MNSFNYTTPDYGHYDDKDTLDLNTPVDKTSNTLRVPDILALVIFAVVFLVGVLGNALVVWVTAFEAKRTINAIWFLNLAVADFLSCLALPILFTSIVQHHHWPFGGAACSILPSLILLNMYASILLLATISADRFLLVFKPIWCQNFRGAGLAWIACAVAWGLALLLTIPSFLYRVVREEYFPPKVLCGVDYSHDKRRERAVAIVRLVLGFLWPLLTLTICYTFILLRTWSRRATRSTKTLKVVVAVVASFFIFWLPYQVTGIMMSFLEPSSPTFLLLNKLDSLCVSFAYINCCINPIIYVVAGQGFQGRLRKSLPSLLRNVLTEESVVRESKSFTRSTVDTMAQKTQAVSpecifications
| C5R1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC008982 | |
| C5AR1 | |
| C5R1 (AAH08982, 1 a.a. ∼ 350 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 4E2 | |
| Mouse monoclonal antibody raised against a full-length recombinant C5R1. | |
| C5AR1 | |
| C5A/C5AR/C5R1/CD88 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 728 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction