Learn More
BrdU (Bromodeoxyuridine) Mouse, Unlabeled, Clone: B44, BD
Description
Anti-BrdU, clone B44, is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with spleen cells from BALB/c mice immunized with iodouridine-conjugated ovalbumin.
Bromodeoxyuridine (BrdU) is a uridine derivative that can be incorporated into DNA in place of thymidine. Anti-BrdU identifies BrdU (but not thymidine) in single-stranded DNA, free BrdU, or BrdU coupled to a protein carrier. The antibody also reacts with iodouridine.
Specifications
Specifications
| Antigen | BrdU (Bromodeoxyuridine) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B44 |
| Concentration | 25μg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Immunogen | Iodouridine |
| Purification Method | Affinity Purified |
| Show More |