Learn More
BDH1 Monoclonal Antibody (1A5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515594
Additional Details : Weight : 0.01000kg
Description
MA5-15594 targets BDH1 in indirect ELISA, IHC and WB applications and shows reactivity with Human and mouse samples. The MA5-15594 immunogen is purified recombinant fragment of human BDH1 expressed in E. Coli. MA5-15594 detects BDH1 which has a predicted molecular weight of approximately 38kDa.
BDH1 Monoclonal antibody specifically detects BDH1 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
BDH1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q02338, Q80XN0 | |
BDH1 | |
Purified recombinant fragment of human BDH1 expressed in E. Coli. | |
RUO | |
622, 71911 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1A5 | |
Unconjugated | |
BDH1 | |
(R)-3-hydroxybutyrate dehydrogenase; 2310032J20Rik; 3-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase (heart, mitochondrial); 3-hydroxybutyrate dehydrogenase, type 1; AI327223; Bdh; Bdh1; b-hydroxybutyrate dehydrogenase; D-beta-hydroxybutyrate dehydrogenase, mitochondrial; MGC2723; MGC4347; MGC9788; SDR9C1; Short chain dehydrogenase/reductase family 9C member 1; short chain dehydrogenase/reductase family 9C, member 1 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |