missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-BCAP31 Matched Antibody Pair, Abnova™
Marca: Abnova H00010134-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-BCAP31 (100μg); Detection antibody: rabbit purified polyclonal anti-BCAP31 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Descrizione
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BCAP31
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair
Specifica
| BCAP31 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BCAP31. | |
| 6C6-AG/BAP31/CDM/DXS1357E | |
| Mouse, Rabbit | |
| RUO | |
| 10134 | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| BCAP31 | |
| BCAP31 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-BCAP31 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-BCAP31 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto