Learn More
anti-APOA2, Human, Matched Antibody Pair, Abnova™
Description
Includes
Capture antibody: mouse monoclonal anti-APOA2 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-APOA2, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
Specifications
| Antigen | APOA2 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Biotin |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human APOA2. |
| Gene | APOA2 |
| Gene Symbols | APOA2 |
| Host Species | Mouse |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Show More |