Learn More
abhydrolase domain containing 5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ABHD5.
Brand: Abnova H00051099-A02.50uL
Description
The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation. [provided by RefSeq
Sequence: FEDDTVTEYIYHCNVQTPSGETAFKNMTIPYGWAKRPMLQRIGKMHPDIPVSVIFGARSCIDGNSGTSIQSLRPHSYVKTIAILGAGHYVYADQPEEFNQKVKSpecifications
| abhydrolase domain containing 5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ABHD5. | |
| ABHD5 | |
| CDS/CGI58/IECN2/MGC8731/NCIE2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 51099 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016006 | |
| ABHD5 | |
| ABHD5 (NP_057090, 240 a.a. ∼ 342 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.