missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANT Polyclonal Antibody
Description
Positive Samples: SW620, BT-474, OVCAR-3, HepG2, Mouse brain; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein Immunogen sequence: VGKGAAQREF HGLGD.
ANT1 catalyzes the exchange of ADP and ATP across the mitochondrial inner membrane.
Specifications
Specifications
| Antigen | ANT |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.06 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC25A4 |
| Gene Accession No. | P12235, P48962, Q05962 |
| Gene Alias | 30 kDa calsequestrin-binding protein; 30 kDa CSQ-binding protein; AAC1; adenine nucleotide translocase-1; adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; ADP ATP translocase; ADP,ATP carrier protein 1; ADP,ATP carrier protein, heart; ADP,ATP carrier protein, heart isoform T1; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP carrier; ADP/ATP translocase; ADP/ATP translocase 1; ADP/ATP translocase T1; ADP/ATP-carrier protein; Anc1; ANT; ANT 1; ANT1; AU019225; CSQ-binding 30 kDa protein; fa22e07; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; MTDPS12; PEO2; PEO3; PEOA2; Slc25a4; SLC25A5; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; solute carrier family 25 member 4; T1; translocase; Unknown (protein for MGC:77591); wu:fa22e07; zgc:77591 |
| Gene Symbols | SLC25A4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?