Learn More
Ansell HyFlex™ 11-501 Aramid Fiber Stretch Armor Gloves, Nitrile Coated
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
Description
- Intercept Technology™ raises the protection & dexterity standards for cut-resistant gloves
- Aramid fiber provides an additional level of protection
- Ergonomic design provides bare-hand closeness and reduces hand fatigue
- Nitrile foam coating enhances grip capabilities and tactile sensitivity
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 3 |
| Coating Material | Nitrile |
| Color | Blue |
| Cut Resistance Rating | ANSI A5 |
| Hand | Specific |
| Latex Free | Yes |
| Material | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | 8 |
| For Use With (Application) | Automotive Repairs, Machinery/Equipment Operation, Metal Fabrication, and Chemical Handling |
| Show More |