missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin V Rabbit anti-Human, Mouse, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11060-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.Specifications
| Annexin V | |
| Polyclonal | |
| Unconjugated | |
| ANXA5 | |
| Anchorin CII, Annexin 5, annexin A5, Annexin V, ANX5, ANXA5, Calphobindin I, CBP I, Endonexin II, ENX2, Lipocortin V, PAP I, PP4, Thromboplastin inhibitor, VAC alpha, Vascular anticoagulant alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100521982, 11747, 308 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry, Immunoprecipitation | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P08758, P48036 | |
| ANXA5 | |
| Annexin V Fusion Protein Ag1538 | |
| 20 μL | |
| Primary | |
| Mouse, Pig, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction