missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Angiotensinogen (1-14), Porcine
For cellular and molecular biology applications
53.49 € - 372.00 €
Specifications
Species | Porcine |
---|---|
Purification Method | HPLC |
Content And Storage | -30°C to -10°C |
Format | Lyophilized Solid |
Molecular Weight (g/mol) | 1759.04 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15427519
|
Thermo Scientific Chemicals
J66239.#0 |
5 mg |
372.00 €
5mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15437519
|
Thermo Scientific™
J66239.M^ |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Porcine | |
-30°C to -10°C | |
1759.04 | |
RUO - research use only |
HPLC | |
Lyophilized Solid | |
Angiotensinogen (1-14), porcine | |
Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser |
RUO – Research Use Only