missing translation for 'onlineSavingsMsg'
Learn More

Androgen R/NR3C4 Antibody (rDHTR/8818), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18580039 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18580039 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18580039 Supplier Novus Biologicals Supplier No. NBP324270JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Androgen R/NR3C4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rDHTR/8818
Conjugate Janelia Fluor 549
Formulation 50 mM Sodium Borate
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Host Species Mouse
Immunogen Recombinant full-length human Androgen R/NR3C4 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 367
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.