missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anamorelin, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
Anamorelin (RC-1291) is a potent ghrelin receptor agonist with EC50 value of 0.74 nM in the FLIPR assay.
Specifications
Specifications
| Chemical Name or Material | Anamorelin |
| CAS | 249921-19-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H42N6O3 |
| Quantity | 5 mg |
| Synonym | RC-1291 ONO-7643 |
| Solubility Information | DMSO : ≥ 100 mg/mL (182.92 mM) |
| SMILES | [H][C@](CC1=CNC2=CC=CC=C21)(NC(C(C)(C)N)=O)C(N3CCC[C@@](C(N(C)N(C)C)=O)(CC4=CC=CC=C4)C3)=O |
| Molecular Weight (g/mol) | 546.7 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction