missing translation for 'onlineSavingsMsg'
Learn More

Anagrelide hydrochloride, MedChemExpress

Product Code. p-200099990
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16428356

Brand: Medchem Express HYB0523A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Anagrelide hydrochloride (BL4162A) is a drug used for the treatment of essential thrombocytosis.

Anagrelide hydrochloride (BL4162A) is a drug used for the treatment of essential thrombocytosis.

Specifications

Chemical Name or Material Anagrelide hydrochloride
Physical Form Solid
CAS 58579-51-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H8Cl3N3O
For Use With (Application) COVID-19-immunoregulation
Synonym BL4162A
Solubility Information DMSO : 7.69 mg/mL (26.29 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES ClC1=C(CN(C2)C3=NC2=O)C(N3)=CC=C1Cl.Cl
Molecular Weight (g/mol) 292.55
Formula Weight 292.55
Percent Purity 99.66%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.