Learn More
Thermo Scientific™ AnaeroPouch™-Bag
Click to view available options
Quantity:
20/Pk.
Unit Size:
Pack of 25
Description
Mitsubishi™ AnaeroPouch-Bag is used to hold AnaeroPouch-Anaero, AnaeroPouch-MicroAero, and AnaeroPouch-CO2 gas generators.
Specifications
Specifications
| Type | AnaeroPouch-Bag |
| Format | Bag |
| Product Line | Thermo Scientific Mitsubishi |
| Quantity | 20/Pk. |