Learn More
Thermo Scientific Chemicals Amyloid beta (1-42), Hydrochloride
Product Code. p-5384488
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Product Code. 15477999
Brand: Thermo Scientific Chemicals J66591.!D
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 4550.49
Product Type Amyloid beta (1-42), hydrochloride
Quantity 0.5 mg
Regulatory Status RUO - research use only