Learn More
Amylase alpha Mouse anti-Human, Porcine, Rat, Clone: 2F1G8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66133-1-IG-20UL
 View more versions of this product
Product Code. 16837735
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
This antibody can recognize amylase alpha.
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.
Specifications
Specifications
Amylase alpha
Monoclonal
0.79 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04746
AMY2A, Amy2a3
Amylase, alpha Fusion Protein Ag8819
20 μL
Primary
Pig, Human, Rat
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
2F1G8
Unconjugated
AMY2A
AMY2A, Amylase, alpha, Pancreatic alpha amylase
Mouse
Protein A
RUO
100521789, 279, 497039
-20°C