Learn More
Tocris Bioscience™ AMTB hydrochloride
GREENER_CHOICE
Product Code. 18046052
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18046052 10 mg 10mg
18025742 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18046052 Supplier Tocris Bioscience™ Supplier No. 3989/10
Please to purchase this item. Need a web account? Register with us today!
TRPM8 blocker
TRPM8 channel blocker. Inhibits icilin -induced TRPM8 channel activation in a rat model (pIC50 = 6.23).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 926023-82-7
Molecular Formula C23H27ClN2O2S
Specifications
Quantity 10 mg
Formula Weight 430.99
Purity >99%
Molecular Weight (g/mol) 430.991
Chemical Name or Material AMTB hydrochloride