Learn More
Beckman Coulter AMPure XP Beads for DNA Cleanup
Product Code. 31723106 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Capacity:
5 mL
60 mL
450 mL
Unit Size:
450mL
5mL
60mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
31723106 60 mL 60mL
31723140 5 mL 5mL
31723123 450 mL 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Refer to SDS
Product Code. 31723106 Supplier Beckman Coulter Supplier No. A63881
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Refer to SDS
The AMPure XP reagent, part of our SPRI reagent family, is a highly efficient, easily automated PCR purification system that delivers superior quality DNA with no salt carryover.
  • Requiring no centrifugation or filtration, AMPure XP reagents can be easily used in manual and automated 96- or 384-well formats.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 60 mL
Isolation Technology Magnetic Bead